Learn More
Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min, Thermo Scientific Chemicals
171.86 € - 637.17 €
Chemical Identifiers
CAS | 12137-21-2 |
---|---|
Molecular Formula | H2O3Pt |
Molecular Weight (g/mol) | 245.10 |
MDL Number | MFCD00066964 |
InChI Key | SPGAMGILENUIOF-UHFFFAOYSA-N |
Synonym | dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2oShow More |
PubChem CID | 11139377 |
IUPAC Name | dioxoplatinum;hydrate |
SMILES | O.O=[Pt]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311629
|
Thermo Scientific Alfa Aesar
012278.03 |
1 g |
171.86 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301629
|
Thermo Scientific Alfa Aesar
012278.06 |
5 g |
637.17 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groupsPlatinum(IV) oxide monohydrate is used as a catalyst in hydrogenation, hydrogenolysis, dehydrogenation, and oxidation reactions in organic synthesis. It also finds application in ceramic industry, inorganic chemistry and in chemical synthesis. It is utilized for the selective reduction of alkenes without reducing nitro group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Hydrogenation catalyst
- Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups
Chemical Identifiers
12137-21-2 | |
245.10 | |
SPGAMGILENUIOF-UHFFFAOYSA-N | |
11139377 | |
O.O=[Pt]=O |
H2O3Pt | |
MFCD00066964 | |
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2oShow More | |
dioxoplatinum;hydrate |
Specifications
450°C | |
Black to Brown | |
H2O3Pt | |
MFCD00066964 | |
14,7527 | |
Insoluble in water,acid,aqua regiaSoluble in water and caustic potash solution; Insoluble in alcohol,acid and aqua regia. | |
O.O=[Pt]=O | |
245.10 | |
245.10 (227.09 Anhydrous) | |
Platinum(IV) oxide monohydrate, Adams Catalyst |
12137-21-2 | |
Powder | |
PtO2·H2O | |
UN1479 | |
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o | |
SPGAMGILENUIOF-UHFFFAOYSA-N | |
dioxoplatinum;hydrate | |
11139377 | |
Pt 75% min. |
Safety and Handling
GHS H Statement
H272
May intensify fire
oxidizer.
P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c
H271
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: I
missing translation for 'einecsNumber' : 215-223-0
missing translation for 'rtecsNumber' : TP2506020
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.