missing translation for 'onlineSavingsMsg'
Learn More

Platinum, nominally 40%, Ruthenium, nominally 20% on carbon black, HiSPEC™ 10000, Thermo Scientific Chemicals

Product Code. 11359668
Click to view available options
Quantity:
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Platinum, nominally 40%, Ruthenium, nominally 20% on carbon black acts as a catalyst used on anodes and cathodes in proton exchange membrane fuel cells (PEMFC) and direct methanol fuel cells (DMFC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
MDL Number MFCD00798552

Specifications

Color Black
Quantity 2 g
UN Number UN1325
Physical Form Powder
Chemical Name or Material Platinum, Nominally 40%, Ruthenium, nominally 20% on carbon black, HiSPEC™ 10000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.