missing translation for 'onlineSavingsMsg'
Learn More

Platinum, nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support, HiSPEC™ 12100, Thermo Scientific™

Product Code. 15423935
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
MDL Number MFCD00798552

Specifications

Color Gray to Black
Quantity 1 g
UN Number UN1325
Chemical Name or Material Platinum, Nominally 50%, Ruthenium nominally 25% on high surface area advanced carbon support, HiSPEC™ 12100
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'