Learn More
Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.
POA - 8279.00 €
Specifications
Chemical Name or Material | Platinum Rhodium foil |
---|---|
Molecular Formula | Pt:Rh; 80:20 wt% |
MDL Number | MFCD00198186 |
Solubility Information | Insoluble in water. |
Percent Purity | 99.9% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384767
|
Thermo Scientific Alfa Aesar
041811.FF |
25 x 25 mm | |||||||
11365939
|
Thermo Scientific Alfa Aesar
041811.FI |
50 x 50 mm |
2584.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374767
|
Thermo Scientific Alfa Aesar
041811.GH |
100 x 100 mm |
8279.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum Rhodium foil is used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Rhodium foil | |
MFCD00198186 | |
99.9% |
Pt:Rh; 80:20 wt% | |
Insoluble in water. | |
Tube |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only