missing translation for 'onlineSavingsMsg'
Learn More

Platinum Rhodium foil, 0.05mm (0.002 in.) thick, 99.9% (metals basis), Thermo Scientific Chemicals

POA - 4868.22 €

Specifications

Chemical Name or Material Platinum Rhodium foil
Name Note 0.05mm (0.002 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 90:10 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD00198186
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354767
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041809.FF
25 x 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11345939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041809.FI
50 x 50 mm
4868.22 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Platinum Rhodium foil, 0.05mm (0.002in) thick, 99.9% (metals basis) is used in the glass industry for furnace linings and complex fabrications. It is also used as a plating material. Further, it acts as a catalyst and as a standard for calibration of base metal thermocouples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum Rhodium foil
Pt:Rh; 90:10 wt%
MFCD00198186
99.9%
≈0.62g/25x25mm
0.05mm (0.002 in.) thick
(metals basis)
Insoluble in water.
Odorless
Foil
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only