missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Rhodium foil, 0.1mm (0.004in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041810.FF
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Rhodium foil | |
Pt:Rh; 90:10 wt% | |
MFCD00198186 | |
99.9% | |
≈1.25g/25x25mm |
0.1mm (0.004 in.) thick | |
(metals basis) | |
25 x 25 mm | |
Odorless | |
Foil |
RUO – Research Use Only