missing translation for 'onlineSavingsMsg'
Learn More

Platinum Wire, 0.076mm (0.003 in.) dia., Thermo Scientific™

Product Code. 11346777
Click to view available options
Quantity:
2 m
50 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: Approx .0.045g/50cm

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
Molecular Formula Pt:Rh; 70:30 wt%
MDL Number MFCD00198186

Specifications

Color Silver
Appearance Silver colored wire
Quantity 50 cm
Odor Odorless
Form Wire
Chemical Name or Material Platinum, 30 wt% Rhodium wire
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.