missing translation for 'onlineSavingsMsg'
Learn More
Platinum wire, 0.368mm (0.0145in) dia, hard, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11312128

Platinum wire, 0.368mm (0.0145in) dia, hard, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11312128
100 cm
Click to view available options
Quantity:
25 cm
100 cm
500 cm
This item is not returnable. View return policy

Product Code. 11312128

Brand: Thermo Scientific Alfa Aesar 013039.BY

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 100 cm
Percent Purity 99.95%
Odor Odorless
Weight ≈2.28g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Platinum wire, 0.368mm (0.0145 in.) dia., Hard, Temper: as drawn

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR17K007 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number013039.BU,013039.CC,013039.BY
Lot NumberY21J008 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number013039.BU,013039.CC,013039.BY
Lot NumberN16J028 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number013039.BY
Lot NumberQ26I013 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog Number013039.BU,013039.CC,013039.BY
Lot NumberI20L21 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013039.BU,013039.CC,013039.BY

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum wire, 0.368mm (0.0145in) dia, hard, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.