missing translation for 'onlineSavingsMsg'
Learn More
Platinum wire, 0.404mm (0.0159in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15426705

Platinum wire, 0.404mm (0.0159in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15426705
50 cm
Click to view available options
Quantity:
10 cm
50 cm
250 cm
This item is not returnable. View return policy

Product Code. 15426705

Brand: Thermo Scientific Alfa Aesar 045058.BW

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Appearance Silvery white wire
Density 21.45g/cm3 at 20°C
Assay Percent Range 99.9% (Metals basis)
Quantity 50 cm
Odor Odorless
Form Wire
Chemical Name or Material Platinum

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN11L045 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog Number045058.CB,045058.BS,045058.BW
Lot NumberT09K008 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog Number045058.CB,045058.BS,045058.BW
Lot NumberR07K019 Certificate TypeCertificate of Analysis Date15/05/2024 Catalog Number045058.CB,045058.BS,045058.BW
Lot NumberP18H012 Certificate TypeCertificate of Analysis Date25/10/2022 Catalog Number045058.CB,045058.BS,045058.BW,045061.06
Lot NumberS08I034 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog Number045058.CB,045058.BS,045058.BW,045061.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum wire, 0.404mm (0.0159in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.