missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 2.0mm (0.08in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11321368
Click to view available options
Quantity:
1 cm
5 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 5 cm
Percent Purity 99.95%
Odor Odorless
Weight ≈67.4g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Platinum wire, 2.0mm (0.0 8 in.) dia., Annealed, Temper: soft

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'