Learn More
BTX™ Pliers for High-Throughout Electroporation Microplate Handler
Pliers for High-Throughout Electroporation Microplate Handler is an accessory for moving or replacing gold contact pins.
Brand:  BTX™ 5012017
Product Code. 15791088
  • 75.25 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Features and Benefits:
  • Made with a specific plasticized tooth to grip the gold contact pins, but not indent or damage the integrity of the voltage conductivity, these pliers come as standard with High-Throughput plate handler, but can also be bought separately.
Specifications
Specifications
BTX High Throughput Electroporation Plate Handler
General Precision
Not Autoclavable