Learn More
Memmert™ Plug-In Tube Extension (Od/Id 60.3/57 mm), Straight, For Exhaust Air Ducting For S75
Use these tube extensions with Memmert ovens. Memmert™ Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.
Brand: Memmert™ S75B29718
This item is not returnable.
View return policy
Description
- Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
- Angled
- Only models U, I, S
Specifications
Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 1060 | |
Plug-In Tube Extension | |
57 x 60.3 mm (I.D. x O.D.) |