missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Plunger Can Sheet
The Plunger Can Sheet is ideal for safely moistening cleaning rags and sponges.
68.00 € - 158.00 €
Product Code | Brand | Material | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Capacity (Metric) | Price | Quantity & Availability | ||||
17866576
|
asecos™
23133 |
Steel | 1 L |
68.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
||||
17816336
|
asecos™
23134 |
Steel | 2.5 L |
79.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
||||
17866536
|
asecos™
23135 |
Stainless Steel | 1 L | N/A | |||||
17816386
|
asecos™
23136 |
Stainless Steel | 2 L |
158.00 €
Each |
Estimated Shipment: 03-03-2025
Log in to see stock. |
||||
Description
- Incl. multilingual safety sticker for content labelling
- Pumping unit with spring-mounted stainless steel dasher plate and integrated flame arrester
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title