Learn More
Thermo Scientific™ PluriTest™ Assay Service
PluriTest Assay Service is a fast and cost-effective alternative to teratoma formation for verification of pluripotency in human pluripotent stem cell lines.
Brand:  Thermo Scientific™ A38154
Product Code. 18745405
  • 354.00 € / Each
Explore more special offers
Description
Description
  • A fast and cost-effective alternative to teratoma formation for verification of pluripotency in human pluripotent stem cell lines
  • This service leverages the GeneChip PrimeView Global Gene Expression Profile Array (905400), which enables expression profiling with an emphasis on established well-annotated content
  • More than 36,000 transcripts and variants are measured per sample
The PluriTest Assay Service offers:
  • Fast data delivery – turnaround time of four weeks or less
  • Convenient sample prep – just send a frozen cell pellet
  • Quantitative results – compare profiles to reference set of >450 cell lines and tissue types
  • Peer endorsement – assay cited >570 times in the scientific literature
  • Exceptional support – a dedicated project manager for frequent project communication
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 sample
Assay Service
Research Use Only
Human
3 Weeks
Applied Biosystems™
Stem Cell Researchers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ PluriTest™ Assay Service > 1 Sample