Learn More
Corning™ Pluronics Sacrificial Ink for Matribot™ Bioprinter
Pluronics Sacrificial Ink is a ready-to-print sacrificial and support ink that exhibits excellent printability. Designed for use with Matribot™ Bioprinter.
Brand:  Corning™ 6157
Product Code. 17760789
  • 97.15 € / Pack of 5
Explore more special offers
Description
Description
  • Ready-to-print sacrificial and support ink that exhibits excellent printability
  • Easy to remove once printed by washing with cold PBS
  • Has been used for the fabrication of vascularized tissues, channels in microfluidic devices, and supporting scaffolds for complicated tissue constructs
  • Can also be diluted to 25% v/v with water to use as a learning tool to mimic dispensing of temperature-sensitive hydrogels
  • Designed for use with Corning™ Matribot™ Bioprinter
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Transparent
    Sacrificial Ink
    Sterile-A Quality
    Fabrication of vascularized tissues, channels in microfluidic devices, and supporting scaffolds for complicated tissue constructs
    4°C to 8°C
    Corning Matribot Bioprinter
    2.7 mL/Syringe
    Pluronics
    5 Syringes
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Corning™ Pluronics Sacrificial Ink for Matribot™ Bioprinter > Pluronics Sacrificial Ink; Quantity: 5 Syringes