missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXNB2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547721
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A2, recombinant human (rh) Plexin A4, rhPlexin B1, rhPlexin B3, rhPlexin C1 and rhPlexin D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
PLXNB2 Polyclonal antibody specifically detects PLXNB2 in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
PLXNB2 | |
Polyclonal | |
Unconjugated | |
PLXNB2 | |
Debt; differentially expressed in brain tumors; dJ402G11.3; KIAA0315; MM1; MM1KIAA0315dJ402G11.3; Nbla00445; PLEXB2; plexin B2; plexin-B2; PLXNB2 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
23654 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O15031 | |
PLXNB2 | |
CHO-derived recombinant human Plexin B2 Leu20-Arg1160 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction