Learn More
Grant Instruments™ PHMT Thermoshaker for Microtubes and Microplates
Accelerates in 3 seconds for fast start but also operates smoothly and quietly, this Grant Instruments™ PMHT Thermoshaker for Microtubes and Microplates is ideal for DNA extraction, enzyme reactions and cellular biology techniques.
Brand: Grant Instruments™ PHMT-PSC18
1534.04 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Combines three instruments in one:
- Microtube and microplate thermoshaker
- Compact benchtop incubator
- Microtube and microplate shaker in cold or ambient temperatures
- 2-line LCD display clearly indicates both set and actual values for temperature, shaking speed and time.
- Interchangeable block for 20 or 12 microtubes provides flexibility for an easy change in application.
- Temperature calibration function to compensate for differences in the thermal behaviour of different tubes and samples.
- Operates extremely smoothly, quietly and consistently.
- Includes ‘soft’ start function for delicate samples.
- Fast start - acceleration 3 seconds.
- Compact and sturdy, with a low profile and small footprint.
Specifications
2 mm | |
Ambient +5° to 100°C | |
0.5 mL and 1.5 mL | |
0.1°C at 37°C; 0.2°C at 60°C; 0.2°C at 100°C | |
130 mm | |
4 kg | |
2 line x 16 charactger LCD |
250 to 1400 rpm (increments 10 rpm) | |
32 x 0.5 mL/1.5 mL Microtubes | |
5°C to 100°C | |
230 mm | |
205 mm | |
PMHT-PSC18 |