Learn More
Brand™ PMMA Macro Cuvettes
Standard macro and micro cuvettes with a clear, clean optical path that indicates the optical path orientation.
Brand:  Brand™ 759105
11.91 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Product Code. 10429852
  • 21.10 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Macro and semi micro cuvettes
  • Sorted by mould cavity number
  • PMMA or PS
  • Minimal extinction value variation
  • Optically perfect transmission range
  • Recessed window to protect against scratches
  • Arrow head marking shows the direction of transmission  
  • Clear, clean optical path with indication of optical path orientation
  • Manufactured under controlled room conditions and packaged fully automatically, without human contact
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Macro Cuvette
2.5 to 4.5 mL
PMMA
100Pack