missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PMP Class A Measuring Flask
PMP flask with polypropylene stopper
Brand: BRAND™ 36180
49.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
500 mL | |
PP stopper | |
0.25 mL | |
A |
PMP | |
Ground joint: NS 19/26 | |
Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction