missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Pneumatic Airgo Crimper

One hand, secure, reproducible crimps/decrimps of 8, 11, 13 and 20mm vials with the push of a button.

Brand:  Fisherbrand™ 00 00 3080

1477.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 15592290

  • 1555.00 € / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 11mm AIRGO Crimper Low Pressure
  • Pneumatic
  • Steel
  • Crimper
  • Reduces hand strain compared to manual crimper operation
  • Quick and easy removal of aluminum seals with the push of a button
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pneumatic Crimper
Steel
High Pressure
11 mm
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers