Learn More
Gibco™ PNGase F Glycan Cleavage Kit
PNGase F Glycan Cleavage Kit includes all components necessary to perform the enzymatic removal of almost all N-linked oligosaccharides from glycoproteins.
Brand:  Gibco™ A39245
Product Code. 16347381
  • 304.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Kit includes the PNGase F enzyme, which cleaves N-glycan chains at the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.
Features:
  • ≥95% purity
  • Recombinant PNGase F enzyme with no endoglycosidase F1, F2, or F3
  • Enzyme leaves N-glycan core oligosaccharides intact and suitable for downstream analysis
  • Suitable for use under native or denaturing conditions
Components included in the PNGase F Glycan Cleavage Kit are: 33,000 units of PNGase F (500,000 units/mL) and 1 mL PNGase F 10X buffer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 33,000 units PNGase F enzyme, recombinant (500,000 units/mL), store at -20°C
• 1 mL PNGase F 10X Buffer, store at -20°C
Liquid
PNGase F Glycan Cleavage Kit
Wet Ice
1 kit