Learn More
Thermo Scientific™ PNPP Substrates
Thermo Scientific™ Pierce PNPP Substrates are powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications.
173.00 € - 286.00 €
Specifications
Product Type | ELISA Substrate |
---|---|
Detection Method | Colorimetric |
Substrate Properties | Chemical Substrate |
Substrate Type | Alkaline Phosphatase |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11889260
|
Thermo Scientific™
34045 |
PNPP Powder |
222.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11859270
|
Thermo Scientific™
34047 |
PNPP Tablets |
226.00 €
105 tablets |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10571384
|
Thermo Scientific™
37620 |
PNPP Substrate Kit |
286.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10371394
|
Thermo Scientific™
37621 |
1-Step™ PNPP Substrate Solution |
173.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PNPP (p-Nitrophenyl Phosphate, Disodium Sal) is a widely used substrate for detecting alkaline phosphatase in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm. Pierce PNPP is available in four formats: dry crystalline powder, single-use tablets, a stable substrate solution, and a complete kit containing substrate solution and diethanolamine buffer.
Thermo Scientific Pierce PNPP (p-Nitrophenyl Phosphate, Disodium Salt) is a colorimetric, soluble substrate of alkaline phosphatase for use in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405 nm.
Features of PNPP (p-nitrophenyl phosphate):
• One-component alkaline phosphatase (AP) substrate
• Absorbance maximum of 405 nm
• Develops a yellow hue in solution upon reaction with AP
• Safer to use—contains a non-mercury, non-azide preservative
The substrate is offered in several formats:
PNPP Substrate
• Powder Form
PNPP Substrate Tablets
• 5 mg tablets with an inert filler as the balance
PNPP Substrate Kit
• PNPP tablets and a Diethanolamine Buffer to yield more than one liter of substrate. The Diethanolamine Substrate Buffer (Cat. No. 34064) is also provided individually as a 5X concentrate.
1-Step™ PNPP Substrate Solution
• Circumvents this time-consuming and variable introducing step by providing a single component PNPP substrate. This substrate is stable for 12 months at 2–8°C and can be stopped with conventional methods.
Specifications
ELISA Substrate | |
Colorimetric | |
Chemical Substrate | |
Alkaline Phosphatase |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.