Learn More
Sampling Systems™ Pocket Sampler
Quick and easy to use sampler that has a GMP design, which means it can be stripped and cleaned very quickly. Sampling Systems™ Pocket Sampler is suitable for taking discrete, zone samples from free flowing powders and granules.
331.00 € - 1955.00 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Handle Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Handle Material | Price | Quantity & Availability | ||
15809241
|
Sampling Systems™
1220J-2000 |
90 mL | 40 mm | 2000 mm | Stainless Steel Inner Rod |
1955.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15819241
|
Sampling Systems™
1220J-900 |
90 mL | 40 mm | 900 mm | Stainless Steel Inner Rod |
1225.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15839231
|
Sampling Systems™
1220A-1500 |
20 mL | 25 mm | 1500 mm | PTFE Inner Rod |
640.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15849231
|
Sampling Systems™
1220A-600 |
15 mL | 25 mm | 600 mm | PTFE Inner Rod |
433.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15859231
|
Sampling Systems™
1220A-900 |
20 mL | 25 mm | 900 mm | PTFE Inner Rod |
441.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15869231
|
Sampling Systems™
1220B-1500 |
20 mL | 25 mm | 1500 mm | Stainless Steel Inner Rod |
785.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15879231
|
Sampling Systems™
1220B-300 |
10 mL | 25 mm | 300 mm | Stainless Steel Inner Rod |
331.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15889231
|
Sampling Systems™
1220B-600 |
15 mL | 25 mm | 600 mm | Stainless Steel Inner Rod |
443.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15899231
|
Sampling Systems™
1220J-1500 |
90 mL | 40 mm | 1500 mm | Stainless Steel Inner Rod |
1525.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
Description
- Made from high quality 316L stainless steel with PTFE or 316L Stainless Steel inner core
- For taking multi-level, discrete samples
- Suitable for free flowing powders
- Bayonet handle that allows for simple operation
- Sealing sleeve enables one pocket to be emptied at a time
- Samplers are supplied with pointed tips as standard
Specifications
Sampler | |
Stainless Steel (316L) |