missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PocketTips™ Pipette Tips

Product Code. 11662020
Click to view available options
Volume (Metric):
100 nL
250 nL
50 nL
500 nL
For Use With (Equipment):
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP96
Hamilton MICROLAB STAR
Molecular Devices FLIPR 384-well Head
Packard 384-well, PerkinElmer
Packard 96-well, PerkinElmer
Unit Size:
Pack of 19200
Pack of 4800
This item is not returnable. View return policy

Product Code. 11662020

Brand: Thermo Scientific™ PE384P30100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Eliminate time-consuming steps, reduce lab ware usage and accelerate assays with PocketTips™ Pipette Tips.

Eliminate time-consuming pipetting steps, reduce lab ware usage and accelerate assays such as PCR primer dilutions with user-friendly PocketTips™ Pipette Tips, which are compatible on multiple workstations. The external dimensions of the tip and rack are the same as standard automation tips, requiring minimal modification and set-up. Simple, economical assay miniaturization preserves compounds and reduces reagent costs.

Features:

  • Transfer samples directly to the assay plate
  • Combine sample and assay material with soft in-tip mixing
  • Coefficient of variation (CV) of less than 6% can be achieved for any pocket size
  • Color: Clear

Recommended for:

Cell-based or low DMSO tolerant assays, selectivity assays (P450), potency determinations (IC50), compound storage and delivery from compound libraries, protein and enzyme aqueous based assays, automated clinical diagnostics and kinetic read (FLIPR) assays.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 nL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Tip Style Automation
For Use With (Equipment) Packard 384-well, PerkinElmer
Product Line PocketTip

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.