Learn More
Thermo Scientific™ PocketTips™ Pipette Tips
Description
Eliminate time-consuming pipetting steps, reduce lab ware usage and accelerate assays such as PCR primer dilutions with user-friendly PocketTips™ Pipette Tips, which are compatible on multiple workstations. The external dimensions of the tip and rack are the same as standard automation tips, requiring minimal modification and set-up. Simple, economical assay miniaturization preserves compounds and reduces reagent costs.
Features:
- Transfer samples directly to the assay plate
- Combine sample and assay material with soft in-tip mixing
- Coefficient of variation (CV) of less than 6% can be achieved for any pocket size
- Color: Clear
Recommended for:
Cell-based or low DMSO tolerant assays, selectivity assays (P450), potency determinations (IC50), compound storage and delivery from compound libraries, protein and enzyme aqueous based assays, automated clinical diagnostics and kinetic read (FLIPR) assays.
Specifications
Specifications
| Volume (Metric) | 250 nL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Format | Rack |
| For Use With (Equipment) | Packard 96-well, PerkinElmer |
| Product Line | PocketTip |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.