missing translation for 'onlineSavingsMsg'
Learn More

Podocarpic acid, MedChemExpress

MedChemExpress Podocarpic acid is a natural product, which has the best all-round positive effect and acts as a novel TRPA1 activator.

POA - 97.20 €

Specifications

Chemical Name or Material Podocarpic acid
CAS 5947-49-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H22O3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16499278
Encompass_Preferred
View Documents
Medchem Express
HY-N2318
10 mg
97.20 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16484937
Encompass_Preferred
View Documents
Medchem Express
HY-N2318
50 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Podocarpic acid is a natural product, which has the best all-round positive effect and acts as a novel TRPA1 activator.
Specifications

Specifications

Podocarpic acid
Off-White
C17H22O3
O=C([C@@]1(C)CCC[C@]2(C)C3=C(CC[C@@]12[H])C=CC(O)=C3)O
274.35
Research
Neuroscience-Neuromodulation
5947-49-9
Research
DMSO : ≥ 100 mg/mL (364.50 mM)
274.35
99.37%
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.