missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Polar Fleece Hat Xtra

Euro Protection™ Polar Fleece Hat Xtra provides comfortable thermal protection for work outdoors and in cold conditions.

Brand:  Euro Protection™ 5COVXN

Product Code. 15814105

  • 337.00 € / Pack of 25
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Good thermal insulation
  • Polar fleece 280g/m2
  • Thinsulate padding 40g/m2
  • Universal fit
Specifications

Specifications

Cold Conditions Hat
Polar Fleece, Thinsulate
280 gsm (Polar Fleece), 40 gsm (Thinsulate)
Universal
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.