Learn More
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles
Confidently store and monitor temperature-sensitive samples
Brand:  Cole-Parmer™ 44260-74
 View more versions of this product
Product Code. 16632874
  • 1855.00 € / Each
Explore more special offers
Description
Description
  • Improve sample protection by maintaining specific temperature ranges for extended time periods
  • Maintain sample integrity with Traceable™ temperature monitoring
  • Data logger is provided with a 2-year NIST-traceable calibration certificate
Specifications
Specifications
35 L
69.85 cm
18.9 in.
48.01 cm
27.5 in.
4.88 in.
12.5 cm
Aluminum