Learn More
3M™ Polisher ST
The Polisher ST is a single-use anion exchange (AEX) product intended to reduce host cell protein, viruses, and other negatively charged contaminants in biopharmaceutical process streams.
Brand:  3M™ 7100228760
Product Code. 17315423
  • 1115.00 € / Pack of 4
Explore more special offers
Description
Description
The 3M™ Polisher ST is intended to reduce host cell protein, viruses, and other negatively charged contaminants in a flow-through polishing chromatography mode in biopharmaceutical process streams. The synthetic, hybrid purifier contains two complementary AEX-functional media: a quaternary ammonium ('Q') functional nonwoven and a guanidinium-functional membrane.
The Q-functional nonwoven provides reduction of turbidity (when present) and DNA. It also provides a portion of the product's AEX capacity for HCP and virus reduction. Guanidinium functionalized membrane downstream of the functionalized nonwoven provides additional AEX polishing capacity. The novel guanidinium functionality of the downstream AEX polishing membrane mimics the amino acid, arginine, one of the three positively charged, naturally occurring standard amino acids that make up proteins and is responsible for viral clearance and high salt tolerance.
Specifications
Specifications
Single Use
Polyamide
1 mL/min.
4
1.4 bar
40°C
4.8 cm
4.3 cm
Polypropylene
1 cm2
Luer Lock
Luer Lock
104°F
1.9 in.
1.7 in.