missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Kimtech™ White Polishing Cloths Refill
Ideal for the fabricated metal manufacturing industry. Kimtech™ White Polishing Cloths Refill are suitable for a variety of dry polishing tasks like surface preparation, pre/post paint applications and polishing glass.
Brand: Kimberly-Clark™ 7212
175.41 EUR valid until 2024-12-31
Use promo code "24047" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for a variety of dry polishing tasks; surface preparation, pre/post paint applications and polishing glass
- For the fabricated metal manufacturing industry
- Spunlace, white cloths
Specifications
Polishing Cloth | |
60 cm | |
39 cm | |
Refill Pack | |
300 Cloths |
23.6 in. | |
15.4 in. | |
White | |
60 x 39 cm | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction