missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 1500, Thermo Scientific Chemicals

Product Code. 10742621
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color White
Physical Form Waxy Flakes
Melting Point 44.0°C to 48.0°C
Linear Formula H(OCH2CH2)nOH
Merck Index 15, 7688
Solubility Information Solubility in water: 650g/L (20°C). Other solubilities: soluble in many organic solvents
Quantity 100 g
Formula Weight 1500
Flash Point >250°C
Hydroxyl Value 70 to 80mg KOH/g
Infrared Spectrum Authentic
Packaging Plastic bottle
Viscosity 36-42 mPa.s (20°C)
Water 0.5% max. (K.F.)
Chemical Name or Material Poly(ethylene glycol), Average M.W. 1500
Show More Show Less
Product Identifier
  • Poly(ethylene glycol), average M.W. 1500

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'