Learn More
MP Biomedicals™ Poly-L-Lysine Hydrochloride
Poly-L-Lysine Hydrochloride is a cellular attachment factor in culture applications.
Brand:  MP Biomedicals™ 0215269090
 View more versions of this product
Product Code. 17348583
  • 1905.00 € / 500mg
Explore more special offers
Description
Description
  • Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates.
  • Poly-L-lysine enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.
  • When adsorbed to the culture surface, poly-L-lysine increases the number of positively charged sites available for cell binding.
  • Polymers of both D- and L-lysine are used to coat solid surfaces.
  • Poly-L-lysine has been reported to improve the protein coating of ELISA plates.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
26124-78-7
-20°C
Cellular attachment factor in culture applications
500 mg
Whitish
Powder
>20,000 g/mol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.