missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl acrylate), solution in toluene, average M.W. ca. 40000 (GPC), Thermo Scientific™

Product Code. 10552471 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 10552471

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 9003-21-8
Physical Form Liquid
Boiling Point 110.0°C
Molecular Formula (C4H6O2)n
Linear Formula [-CH2CH(CO2CH3)-]n
MDL Number MFCD00084441
Density 1.2200g/mL
Synonym 4-phenylmethoxyphenyl methyl prop-2-enoate
Solubility Information Solubility in water: .. Other solubilities: soluble in esters, aromatic hydrocarbons, ketones,
Quantity 500 mg
InChI Key PTMPEQAUGJJXJV-UHFFFAOYSA-N
SMILES COC(=O)C(-*)C-*
IUPAC Name [4-(benzyloxy)phenyl]methyl prop-2-enoate
Molecular Weight (g/mol) 268.31
PubChem CID 4206998
Flash Point 4°C
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.22
Chemical Name or Material Poly(methyl acrylate)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Poly(methyl acrylate), solution in toluene, average M.W. ca. 40000 (GPC)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Toluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.