missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 31,000-50,000, Thermo Scientific Chemicals

Product Code. 15235968
Click to view available options
Quantity:
100 g
1 kg
2.5 kg

Indicator

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Melting Point 230.0°C to 240.0°C
pH 4.5 to 6.5 (4% aq. soln.)
Infrared Spectrum Authentic
Linear Formula [-CH2CH(OH)-]n
Quantity 2.5 kg
Merck Index 15,7706
Solubility Information Solubility in water: soluble in hot water.
Viscosity 5.2-6.2 mPa.s (20°C 4% aq. sol.)
Percent Purity 87.0-89%
Chemical Name or Material Poly(vinyl alcohol)
Product Identifier
  • Poly(vinyl alcohol, 87.0-89.0% hydrolyzed, average M.W. approx. 31000-50000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'