missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145,000 to 180,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 230.0°C to 240.0°C |
Solubility Information | Solubility in water: soluble in hot water. |
Quantity | 1 kg |
Packaging | Plastic bottle |
Chemical Name or Material | Poly(vinyl alcohol) |
Safety and Handling
Product Identifier
- Poly(vinyl alcohol), 88% hydrolyzed, average M.W. 145000-180000
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'