missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt.%, Thermo Scientific Chemicals

Product Code. 10183960
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25702-80-1
Molecular Formula (C2H3R)n
Molecular Weight (g/mol) 134.56
MDL Number MFCD00084460
InChI Key SQNNHEYXAJPPKH-UHFFFAOYSA-N
Synonym 2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride
PubChem CID 3080734
IUPAC Name chloroethene; prop-2-enoic acid
SMILES *-CC(-*)[*]

Specifications

Color White
Physical Form Amorphous Powder
Density 1.3900g/mL
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, carbon disulfide, mek, thf
Quantity 250 g
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.39
Chemical Name or Material Poly(vinyl chloride), carboxylated
Product Identifier
  • Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'