missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt.%, Thermo Scientific Chemicals

Product Code. 10573741
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25702-80-1
Molecular Formula (C2H3R)n
Molecular Weight (g/mol) 134.56
MDL Number MFCD00084460
InChI Key SQNNHEYXAJPPKH-UHFFFAOYSA-N
Synonym 2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride
PubChem CID 3080734
IUPAC Name chloroethene; prop-2-enoic acid
SMILES *-CC(-*)[*]

Specifications

Color White
Physical Form Amorphous Powder
Density 1.3900g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.39
Chemical Name or Material Poly(vinyl chloride), carboxylated
Product Identifier
  • Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.