Learn More
Buerkle™ Polyacetal Hose Clips
Use these Buerkle™ Polyacetal Hose Clips to clamp down on tubing for a sure and secure fit.
5.25 € - 6.50 €
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
10382482
|
Buerkle™
8678-0006 |
6 to 6.5 mm |
6.10 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10054083
|
Buerkle™
8678-0010 |
9 to 10 mm |
5.25 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
10591954
|
Buerkle™
8678-0011 |
10 to 11 mm |
5.25 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10696453
|
Buerkle™
8678-0012 |
11 to 12 mm |
5.25 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10542403
|
Buerkle™
8678-0014 |
12 to 14 mm |
5.25 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10347752
|
Buerkle™
8678-0015 |
13 to 15 mm |
5.25 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
10275872
|
Buerkle™
8678-0017 |
15 to 17 mm |
5.25 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
10163042
|
Buerkle™
8678-0019 |
17 to 19 mm |
5.25 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10616843
|
Buerkle™
8678-0020 |
18 to 20 mm |
6.50 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10001922
|
Buerkle™
8678-0022 |
20 to 22 mm |
6.50 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10033513
|
Buerkle™
8678-0025 |
23 to 25 mm |
6.50 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
- Hose clips made of Polyacetal (POM)
- Featured in varying diameters
- POM is white, sterilizable, does not rust, permanently elastic and hygienic.
Specifications
Polyacetal | |
Hoses | |
Hose Clip |