Learn More
Quickfit™ Polyacetal Joint Clips
Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.
Brand:  Quickfit™ KC10
 View more versions of this product
Product Code. 10488960
  • 52.40 € / Pack of 10
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.
  • Effortless attachment and removal of clips
  • Reduces the need for traditional clamping
  • Withstands temperatures up to 80°C
  • Color coded sizes for quick identification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Polyacetal
Joint Size 10/19
Joint Clamp
Glassware with conical joints
Mint