missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Polyacetal Joint Clips

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

Brand:  Quickfit™ KC45

70.78 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12029072

  • 76.40 € / Pack of 10
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from polyacetal for strength and rigidity. Quickfit™ Polyacetal Clamps for Conical Joints are designed to secure corresponding Quickfit™ glassware with conical joints.

  • Effortless attachment and removal of clips
  • Reduces the need for traditional clamping
  • Withstands temperatures up to 80°C
  • Color coded sizes for quick identification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10/Pk.
Polyacetal
Joint Size 45/40
Joint Clamp
Glassware with conical joints
Brown
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers