Learn More
Koflo Unmounted In-Line Static Mixers
Unmounted In-Line Static Mixers can be used with any standard tubing having the same tubing ID as the element OD given in the table.
Brand:  Koflo 04667-18
 View more versions of this product
Product Code. 11774109
  • 122.00 € / Pack of 10
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • They work well with flexible tubing.
Specifications
Specifications
0.5 in.
10 in.
Unmounted In-line Mixer
13 mm
254 mm