Learn More
Scilabub™ Polyamide Fiber Protective Gloves
These Nomex™ polyamide heat resistant gauntlet gloves are resistant to 260°C (working temperature, dry heat only) with knitted fabric construction for extra comfort.
Brand:  Scilabub™ GAU/NM
 View more versions of this product
Product Code. 12302239
  • 381.00 € / 1 pair
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Heat-resistant (260°C working temperature, dry heat only)
  • Low thermal conductivity
  • Asbestos free
  • Double thickness
  • Knitted fabric for comfort
  • Does not shrink or melt
  • Unaffected by washing
  • Hard wearing
  • Conform to EN 388: 1 X 4 X and EN 407: 4 1 3 0 1 X
Specifications
Specifications
CE Mark
Polyamide
Protective Gloves
Non-sterile
Red
Powder Free
Medium
Heat, Chemical