missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Polyamide Layer Sheets for Chromatography

Polyamide used as a catalyst in the epoxidation of cyclohexene with tert-butylhydroperoxide

184.92 € - 723.00 €

Specifications

Content And Storage Ambient temperatures
Material Polyamide
For Use With (Application) Chromatography
Dimensions (L x W) 15 x 15cm
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459010
View Documents
Thermo Scientific™
A12572.KT
1/ea.
184.92 €
Each
Estimated Shipment: 31-12-2024
to see stock.
Add to basket
 
11469010
View Documents
Thermo Scientific™
A12572.KU
5/ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Polymer-supported reagents and catalysts
  • Easy recovery upon completion of reaction
  • Used for Chromatography
Specifications

Specifications

Ambient temperatures
Polyamide
Chromatography
15 x 15cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.