Learn More
Cytiva Nets for C 10 and AC 10
Resistant to aqueous solutions and solvents used in liquid chromatography.
Brand:  Cytiva 19-5008-01
Product Code. 10839600
  • 53.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with C 10 columns
  • Made from polyamide and polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nets for C 10 and AC 10
Resistant to aqueous solutions and solvents used in liquid chromatography
C 10 Columns
Polyamide, Polypropylene