Learn More
Tocris Bioscience™ Polybrene
Viral transduction enhancer
Brand:  Tocris Bioscience™ 7711/10
91.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18717326
  • 120.00 € / 10mg
Explore more special offers
Description
Description
Polybrene is a cationic polymer that improves lentiviral and adenoviral transduction efficiency in mammalian cells in vitro. It also enhances DNA transfection in many types of cells. It has a neutralizing effect on negatively charged DNA and viral particles which facilitates their attachment and entry into host cells. Polybrene is suitable for use with CRISPR/Cas9 protocols.
Polybrene is a lyophilized solid and can be prepared as a stock solution by adding 1 mL of sterile ultra-pure water directly to the vial (final stock concentration 10 mg/mL).
For further information view the product protocol .
Specifications
Specifications
1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide
10 mg
0.95
28728-55-4
Viral Transduction Enhancers