missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Polycarbonate Dome for FastPrep-24™

Polycarbonate Dome is a protective cover for FastPrep-24™ Classic bead beater.

Brand:  MP Biomedicals™ 116002506

Product Code. 11432370

  • 438.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Applications:
    • Dome for FastPreps-24
    • Polycarbonate protective cover
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polycarbonate
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.