missing translation for 'onlineSavingsMsg'
Learn More

Polycarbonate resin, ∽ M.W. 45.000, pellets, Thermo Scientific Chemicals

Product Code. 10307840
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24936-68-3
Molecular Formula (C16H14O3)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084476
SMILES CC(C)(C1=CC=C(-*)C=C1)C1=CC=C(OC(=O)O-*)C=C1

Specifications

Color Undesignated
Physical Form Pellets
Packaging Glass bottle
Density 1.2000g/mL
Quantity 5 g
Linear Formula [-C6H4C(CH3)2C6H4OCO2-]n
Chemical Name or Material Polycarbonate resin, M.W. 64.000
Product Identifier
  • Polycarbonate resin, ca. M.W. 45.000, pellets

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'