missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Polyester Cable
Use this solid braided line with sampling equipment that weighs less than 75 lb (34 kg)
Brand: Cole-Parmer™ 05499-35
This item is not returnable.
View return policy
Description
- Maximum load is 110 lb (50 kg)
Specifications
Cable | |
Polyester | |
1 Ea. | |
30 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction