missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ Durx™ 670 General Purpose Wipes
BIOPHARMA

Use these wipes for cleaning and maintenance in laboratories as well as aerospace and defense, microelectronics, food processing, pharmaceutical and biomedical device manufacturing environments. Berkshire™ Durx™ 670 General Purpose Wipes feature exceptional strength and absorbency.

Brand:  Berkshire™ DR670.0909.20

33.35 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 12877143

  • 36.00 € / Pack of 300
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured without the use of binders or surfactants, ion levels and organic extractables remain low
  • The same wipe material can be provided in pre-wetted formats for reduced VOC emissions, increased convenience, increased productivity, improved solvent control and cleaning protocol repeatability and reduced costs
  • For aseptic processing areas, the same wipe material can be provided in a gamma irradiated format, validated sterile to a 10–6 sterility assurance level
  • Dry and pre-wetted versions available
  • FDA 21 CFR – Food contact compliant
Specifications

Specifications

Cleanroom Wiper
230 mm
Knife-Cut
230 mm
Polyester Cellulose
300 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.