Learn More
Brady™ Polyester Labels
Polyester Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling.
Brand:  Brady™ 143075
 View more versions of this product
Product Code. 13533320
  • 150.00 € / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, and weather-resistant.
Specifications
Specifications
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognised
Permanent Rubber
White
Polyester
Labels
BBP33, i3300, i5300
3
5000 Labels/Box
Rectangle
-40°F to +248°F
0.004 in.
15.24 mm
0.5 in.
1 in.
Polyester Labels for BBP33/i3300 Printers
Powder
Gloss
Thermal Transfer
Asset tracking, Barcode labelling, Component and equipment labelling, Data and telecommunications labelling, Electrical labelling, General Identification, Inventory and inspection labelling
27.3 mm
-40°C
5000 Pk.
LabelMark, MarkWare
-40°C to +120°C
0.102 mm
12.7 x 25.4 mm
12.7 mm
25.4 mm